Lecithin (from soy bean)

Leverandør: VWR Chemicals

298632AEA 1400 NOK
298632A
Lecithin (from soy bean)
Lecithin
MDL nummer: MFCD00147406
CAS-nummer: 8030-76-0
EINECS: 310-129-7
Merck Indeks: 12,05452

Bestill nå

Spesifikasjon Testresultater

Assay of nitrogen 0.6 - 1.2 %
Assay (Phosphorus) 2.0 - 3.3 %
Sulfated ash (800°C) Max. 10 %
Microbiological test Passes test
Salmonella species (absent in 10 g) Passes test
Escherischia coli (absent in 1 g) Passes test
Enterobacteriaceae (in 1 g) Absent


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR