VWR®, Sampler for frozen foods
Leverandør: VWR Collection
300-0164EA 3400 NOK
300-0164
VWR®, Sampler for frozen foods
Prøvetakere Prøveøser
Cylindrical probe for the proper sampling of frozen products packed in bulk - to be used with electrical drill - suitable for collecting frozen goods. Ideal to use in the food industry and for health authorities. Method of use: Probe is fixed to an electric drill. The drill is activated and then pushed against the frozen surface to be sampled. The sampling is made easy by the anti-clockwise rotation of the drill. The 'carrot' is extracted from the probe with a rod and pushed into the sampling container. All these operations are aseptically performed in case of microbiological analysis.