VWR®, Ice sampler
Leverandør: VWR Collection
300-0165EA 2075 NOK
300-0165
VWR®, Ice sampler
Prøvetakere Prøveøser
Cylindrical probe for aseptic sampling of ice cream and similar products. The probe is completed with a special PYREX® test tube for transport of the sample to the laboratory for the food industry, also ice cream producers and health authorities.
  • AISI 316L stainless steel and chromium plated brass cylindrical probe with handle
  • PYREX® glass test tube
  • Sterilisable
Method of use: The sterilised cylindrical probe is extracted from the test tube and inserted into the sample. The probe is transferred into the test tube and in this condition is delivered toMore Product Information