Sterilin® Dippas® Milk Dipper
Leverandør: STERILIN
194IW
300-0200EA 8575 NOK
300-0200
Sterilin® Dippas® Milk Dipper
Prøvetakere Prøveøser
Clear polystyrene container with snap-off handle and leakproof metal screw cap enabling samples to be sent for analysis without risk of cross-contamination.
  • Individually wrapped and sterilized by gamma irradiation
  • Leak tested in accordance with BS EN ISO 6717
  • Available in either clear or blue polystyrene
  • Handle neatly snaps off after sample collection for ease of transportation
  • Choice of handle length to accommodate the majority of sampling applications
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904