Disposable samplers, MultiDispo
Leverandør: BURKLE
5393-4440 5393-4441 5393-4450 5393-4451
300-0245EA 7000 NOK
300-0245 300-0246 300-0247 300-0248
Disposable samplers, MultiDispo
Prøvetakere Prøvetakere til engangsbruk
This MultiDispo zone samplers for taking samples from multiple layers of the sample material.
  • Easy emptying through the opening (Ø 14 mm) in the handle
  • Clean room manufactured
  • Cross contamination is completely eliminated
Suitable for free-flowing powders and small-grain granulates.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Miljøsertifikat ISO14001 Certificate ISO 14001 Certificate