Isborere
Leverandør: BURKLE
5323-0200 5323-2010 5323-0190
312-0072EA 10125 NOK
312-0072 312-0086 300-0357
Isborere
Prøvetakere Prøvetakere til faststoff
Sampler for deep-frozen products in the food and luxury items sector, and for semi-solid substances. The very robust hollow drill cuts quickly into the sample material with its sharp sawing crown. A sample cylinder (cross-section sample) can be cut out.
  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilisable
  • Cross-section sample
The ice drill can be mounted on a standard commercial drilling machine. The core bit drills quickly and easily down to the desired depth in the frozen material. This makes it possible to cut a representativeMore Product Information