Measuring spoons, blue
Leverandør: BURKLE
5378-4051 5378-4052 5378-4053 5378-4054 5378-4059 5378-4044 5378-4058 5378-4045 5378-4046 5378-4047 5378-4055 5378-4056 5378-4057 5378-4048 5378-4049 5378-4050
300-0386EA 1600 NOK
300-0386 300-0387 300-0388 300-0389 300-0394 300-0379 300-0393 300-0380 300-0381 300-0382 300-0390 300-0391 300-0392 300-0383 300-0384 300-0385
Measuring spoons, blue
Skjeer
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
  • PS, blue
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • With stripper edge
  • For single use
  • Clean room manufactured (clean room class 7)
The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in referenceMore Product Information