Manual sampler, Powder Max
Leverandør: Sampling Systems
1213A-01
300-0431EA 12175 NOK
300-0431
Manual sampler, Powder Max
Prøvetakere Prøvetakere til faststoff
The Powder Max is ideal for taking large samples of free-flowing powders.
  • Takes large samples
  • Can be fully dismantled for easy cleaning
  • Easy operation
This single point sampler is ideal for sampling large volumes of free-flowing powders and granules. The intuitive design and self closing sleeve means the sampler remains closed until it is pulled out. After use, the sampler can be fully dismantled for easy cleaning, in line with GMP guidelines.