Hand paddle with holes, Sampling Systems
Leverandør: Sampling Systems
8026P-540
300-0517EA 1050 NOK
300-0517
Hand paddle with holes, Sampling Systems
Omrørere Omrørerstaver
The hand paddle with holes is perfect for stirring and mixing liquids.
  • Short, ergonomic handle for ease of use
  • Holes allow for easy mixing
  • Mde from robust chemicall resistant PP
  • 133 mm blade width
The hand paddle is excellent for stirring liquids in large containers, and the ergonomic handle allows it to be used with one hand or with both hands.