Ice scoop, stainless steel
Leverandør: Sampling Systems
A314-100
300-0664EA 590 NOK
300-0664
Ice scoop, stainless steel
Øser
The ice scoop is designed for scooping ice cubes and pieces of broken ice.
  • 304 stainless steel
  • Smooth handle
  • Robust design
  • Easy to use
The body of the scoop has many 3 mm perforations to allow any melt water to easily drain away.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904