Sampler, SiloPicker
Leverandør: BURKLE
5340-1100
300-1025EA 14025 NOK
300-1025
Sampler, SiloPicker
Prøvetakere Prøvetakere til faststoff
For sampling bulk goods at depths of up to 3,5 metres, depending on the density of the bulk goods to be sampled. Inserting the sampler into the bulk goods closes the collection chamber. At the desired depth, the collection chamber is opened by withdrawing the sampler, the chamber is filled and can be pulled out and collected.
  • Easy handling
  • Sampler length 1300 mm, may be extended up to 3500 mm using extension rods
  • Stainless steel V4A (1.4404)/AISI 316 L