Prøvetaker for væsker, UniSampler
Leverandør: BURKLE
5314-2000
300-1042EA 6675 NOK
300-1042
Prøvetaker for væsker, UniSampler
Prøvetakere Væskeprøvetakere
Hand operated vacuum pump sampler with a 2,5 metre long PVC hose, the stainless steel weight will sink the end of the hose to the required sampling depth.
  • Deep, narrow, and hard to reach spots
  • Powerful suction for viscous media, easy to disassemble and clean
  • Universal bottle adapter made of PVC for 24, 32, 39 and 50 mm Ø bottle necks and for 100 to 1000 ml sample bottles
Liquid only comes into contact with the suction hose and sample bottle.
Leveringsinformasjon: Supplied in case with handpump, adapter for bottles, sample bottle 1000 ml, PVCMore Product Information