Sampler, UniSampler “Ex”
Leverandør: BURKLE
5314-3000
300-1044EA 12275 NOK
300-1044
Sampler, UniSampler “Ex”
Prøvetakere Væskeprøvetakere
Glass bottle and hose are connected in such a way that they conduct electricity and are grounded via a grounding cable. Operation: Ground the device, immerse the suction hose to the required depth, attach the adapter to the sampling bottle, generate a vacuum with the manual pump and collect the sample, remove the filled sampling bottle, close it and label it.
  • Specially developed for sampling flammable liquids of hazardous materials (e.g. gasoline)
  • PTB approval D- 333-009 618/00
Specially developped for sampling flammable liquids of hazardousMore Product Information