Engangsprøvetakere, LiquiThief, SteriWare®
Leverandør: Sampling Systems
8050H-500 8050H-500S 8050H-1000 8050H-1000S 8050P-1000 8050P-500 8050H-1500 8050H-1500S
300-2002EA 4400 NOK
300-2002 300-2003 300-2000 300-2001 300-2004 300-2005 300-2032 300-0290
Engangsprøvetakere, LiquiThief, SteriWare®
Prøvetakere Prøvetakere til engangsbruk
Disposable liquid samplers designed for a high number of liquids and low viscosity testing applications.
  • No risk of cross- contamination - can be used straight from the bag
  • Silicone seal for improved sampling performance
  • Assembled and packed in a Class 100 000 cleanroom
Designed for taking fast sterile samples from tanks, kegs, and containers in pharmaceutical, food, chemical, cosmetic, and biotechnology industries.
Sertifiseringer: FDA, EU 10/2011 and EC 1935/2004 conformity. BSE-/TSE-free. Full batch traceability, ATEX rated.