Cytoplasmic membrane labelling kits, CellBrite™
Leverandør: Biotium
30021 30022 30023 30024
30021.EA 2900 NOK
30021. 30022. 30023. 30024.
Cytoplasmic membrane labelling kits, CellBrite™
Assay Cellulær analyse
CellBrite kits are designed to label cell cytoplasmic membranes with a fluorescent membrane dye pre-dissolved as an optimised staining solution. By combining CellBrite membrane stains, it is possible to label multiple cell populations with distinctive colours, a capability useful for the studies of cell-cell interactions.
  • Direct addition of the dye to normal culture media followed by a brief incubation period
  • Highly lipophilic dye ensuring no dye transfer between cells, resulting in stable labelling
  • Bright, photostable, and non toxic
The labelling is stable and non toxic, suitable for long-term tracking of cells.