EDTA dinatrium salt 0.0178 mol/l (N/28) in water, AVS TITRINORM for hardness of water determinations, for hardness of water determinations

Leverandør: VWR Chemicals

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

30029.290EA 365 NOK
30029.290
EDTA dinatrium salt 0.0178 mol/l (N/28) in water, AVS TITRINORM for hardness of water determinations, for hardness of water determinations
EDTA dinatrium salt

1 ml = 1°dH/100 ml H₂O

Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Tetthet: 1.001 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå

Spesifikasjon Testresultater

Titer (20 °C; real value 0,2 % accuracy) 0,017764 - 0,01784 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR