Eddiksyre 0.1 mol/l (0.1 N), AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

300652.1000EA 485 NOK
300652.1000
Eddiksyre 0.1 mol/l (0.1 N), AVS TITRINORM volumetric solution
Eddiksyre glasial (iseddik)
Formel: H₃CCOOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR