Perchloric acid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric

Leverandør: VWR Chemicals
Warning

30111.291EA 825 NOK
30111.291
Perchloric acid 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Perchloric acid
Formel: HClO₄
MW: 100.46 g/mol
Kokepunkt: 39
Smeltepunkt: -101
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 3264
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR