Azure B tetrafluoroborate 95% Dye, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium tetrafluoroborate

302619-5G
302619-5GEA 1768.08 NOK
302619-5G
Azure B tetrafluoroborate 95% Dye, Sigma-Aldrich®
Azure B tetrafluoroborate

Azure B tetrafluoroborate, Dye content 95 %

Formel: C₁₅H₁₆BF₄N₃S
MW: 357.18 g/mol
Smeltepunkt: 244 °C (dec., lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011761
CAS-nummer: 79288-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=894b3d4c3b34c42983321afe6c0e48576a47117f&vid=5e55778bf5b974f88053ed784f5fc98dc8541a5f&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR