Ammoniumcerium(IV)nitrat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

30483.295EA 1625 NOK
30483.295
Ammoniumcerium(IV)nitrat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Ammoniumcerium(IV)nitrat
Formel: (NH₄)₂Ce(NO₃)₆
MDL nummer: MFCD00151121
CAS-nummer: 16774-21-3
EINECS: 240-827-6

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR