Benzetoniumklorid 0.004 mol/l, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

30497.293EA 420 NOK
30497.293 30497.327
Benzetoniumklorid 0.004 mol/l, AVS TITRINORM volumetric solution
Benzetoniumklorid
Formel: C₂₇H₄₂ClNO₂
MDL nummer: MFCD00011742
CAS-nummer: 121-54-0
EINECS: 204-479-9

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.00399 - 0.00401 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR