Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

30498.296EA 465 NOK
30498.296
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Zinc sulphate
Formel: O₄SZn
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011302
CAS-nummer: 7733-02-0
EINECS: 231-793-3
FN: 3082
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR