Cerium1000 mg/L Ce in 5% salpetersyre (fra Ce) Specpure® standard til AAS

Leverandør: Thermo Fisher Scientific

30520.AE 30520.AP
30520.AEEA 715 NOK
30520.AE 30520.AP
Cerium1000 mg/L Ce in 5% salpetersyre (fra Ce) Specpure® standard til AAS
Standarder Kationstandarder Cerium Standards

Molecular Formula: Ce in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010924


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR