Acid Blue 129 25% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Atanyl Blue PRL

306495-25G
306495-25GEA 639 NOK
306495-25G
Acid Blue 129 25% (dye content), Sigma-Aldrich®
Acid Blue 129

Acid Blue 129, Dye content 25 % (Atanyl Blue PRL)

Formel: C₂₃H₂₁N₂NaO₅S
MW: 460.48 g/mol
Smeltepunkt: >300 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013263
CAS-nummer: 6397-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d630512150afb7797964556f2948397762c981d6&vid=52a61e140414a8191f689f98bbc501a692f34a1f&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR