Tetrametylammoniumhydroksid 25% (w/w) in methanol

Leverandør: Thermo Fisher Scientific
Danger

30833.09 30833.22 30833.36
30833.09EA 530 NOK
30833.09 30833.22 30833.36
Tetrametylammoniumhydroksid 25% (w/w) in methanol
Tetrametylammoniumhydroksid

CAS No.: 75-59-2 Molecular Formula: C4H13NO Formula Weight: 91.15 Flash Point: 12°(54°F) Physical Form: Liquid Density: 0.865 Appearance: Colourless to pale yellow Odour: Like alcohol Merck Reference: 14,9224 UN No.: 2924 MDL No.: MFCD00008280

Formel: C₄H₁₃NO
Tetthet: 0,865
Flash punkt: 12 °C (54 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008280
CAS-nummer: 75-59-2
EINECS: 200-882-9
FN: 2924
ADR: 3,II
Merck Indeks: 14,9224

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR