Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution

Leverandør: VWR Chemicals

Synonymer: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

30882.294EA 950 NOK
30882.294
Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Ammonium iron(II) sulphate
Formel: H₈FeN₂O₈S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051039
CAS-nummer: 10045-89-3
EINECS: 233-151-8

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR