Schiff reagent for analysis of aldehydes

Leverandør: VWR Chemicals

30969.261EA 750 NOK
30969.261
Schiff reagent for analysis of aldehydes
Schiff reagent

Remark: In the particular case of formaldehyde, use an ethyl alcohol solution so that the final solution contains about 50% of ethyl alcohol.

Bestill nå

Spesifikasjon Testresultater

Sensitivity Passes test
Density (20/4) 1.005 - 1.015


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR