Natriumdodecylsulfat 0.01 mol/l in aqueous solution

Leverandør: VWR Chemicals

Synonymer: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

309747.1000EA 576 NOK
309747.1000
Natriumdodecylsulfat 0.01 mol/l in aqueous solution
Natriumdodecylsulfat
Formel: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Tetthet: 1…1.1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036175
CAS-nummer: 151-21-3
EINECS: 205-788-1
Merck Indeks: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR