Salpetersyre 6 mol/l (6 N), AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Danger

Synonymer: Nitric acid solution

310547.1000EA 645 NOK
310547.1000
Salpetersyre 6 mol/l (6 N), AVS TITRINORM volumetric solution
Salpetersyre
Formel: HNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 6.00 N +/- 0,2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR