Next generation dsDNA quantification kits, AccuBlue®
Leverandør: Biotium
31060 31060-T 31061 31061-T
31060.EA 8325 NOK
31060. 31060-T 31061. 31061-T
Next generation dsDNA quantification kits, AccuBlue®
Nukleinsyre reagenser Next Generation Sequencing Reagents
The AccuBlue® NextGen dsDNA Quantitation Kit offers unprecedented level of sensitivity in dsDNA detection. It is a sensitive and accurate DNA quantitation method for ultra-low amounts of DNA with a detection limit as low as one pg DNA in a 96-well plate reader assay.
  • Detection limit: 1 pg DNA*
  • Linear range: 1 to 3000 pg dsDNA per well of a 96-well plate (0,5 pg/µl to 300 pg/µl of sample)
  • Unrivaled sensitivity
  • Green fluorescent dye with blue LED excitation sources (excitation/emission wavelengths: 468/507 nm)
The AccuBlue® NextGen dsDNA Quantitation Kit provides an unprecedented level of sensitivity in dsDNA detection. It is ideal for use in preparing samples for applications such as Next Generation Sequencing (NGS) and digital PCR. It is designed for use with fluorescence 96-well plate readers equipped with excitation and emission filters for detecting green fluorescence. The unique spectral properties make it well-suited for use with instruments with blue LED excitation sources.
*Limit of detection in the range of 1 to 5 pg depending on instrument sensitivity
Leveringsinformasjon: Kit contains AccuBlue® NextGen dye, 400X; AccuBlue® NextGen enhancer, 100X; AccuBlue® NextGen buffer, 1X; high sensitivity DNA standard, 10 ng/μl. Standalone AccuBlue® quantitation solutions are available for users who prefer to use their own DNA standards.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon