Meyer's reagent for blood research
Leverandør: VWR Chemicals
Warning
31072.268EA 1975 NOK
31072.268
Meyer's reagent for blood research
Meyer's reagent
Kokepunkt: >100 °C (1013 hPa)
Tetthet: 1.1 g/cm³ (20 °C)
FN: 1760
ADR: 8 II