for cellulose dissolution

Leverandør: VWR Chemicals
Warning

31115.267EA 1750 NOK
31115.267
for cellulose dissolution
Schweitzer's reagent (tetraamminecopper(II) dihydroxide solution, shirley's)
Formel: CuH₁₄N₄O₂
MW: 165.68 g/mol
EINECS: 241-508-4
FN: 3266
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR