Supelco® PESTANAL® Organic Reference Standard, Pencycuron

Leverandør: Merck

31118-100MG
31118-100MGEA 810 NOK
31118-100MG
Supelco® PESTANAL® Organic Reference Standard, Pencycuron
Standarder Organiske standarder

NB: P273 - P391 - P501

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10a5e65df0ee7e3c1448831cfeb1d118d6ebee48&vid=b1b8f9239a07638a563054f11f1b4ecd92cf9d0a&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 66063-05-6
MDL No. MFCD00078724
Formel C₁₉H₂₁ClN₂O
Molekylvekt 328,84
Hardhetsklasse beskrivelse H410
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR