Saltsyre 0.01 mol/l (0.01 N), VOLUSOL® Volumetric

Leverandør: VWR Chemicals

Synonymer: Hydrochloric acid solution, Muriatic acid

311341.1000EA 321 NOK
311341.1000
Saltsyre 0.01 mol/l (0.01 N), VOLUSOL® Volumetric
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Tetthet: 1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 III

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.00998 - 0.01002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR