Maursyre ( metansyre) 85%, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

3127-46EA 1225 NOK
3127-46
Maursyre ( metansyre) 85%, AR®, Macron Fine Chemicals™
Maursyre ( metansyre)
Formel: HCO₂H
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003297
CAS-nummer: 64-18-6
FN: 1779
ADR: 8 (3),II

Bestill nå

Spesifikasjon Testresultater

Appearance colorless, clear liquid Passes test
Assay 84 - 86 % 85.6 % 84 - 86 % 85,6 %
Acetic acid (CH3COOH) ≤0,5 %
Oxalic acid (H2C2O4) Passes test
Residue after Evaporation ≤0,003 %
Water precipitated substances Passes test
Chloride (Cl) ≤0,0005 %
Sulphate (SO4) ≤0,003 %
Sulfite (SO3) ≤0,0015 %
Heavy Metals (as Pb) ≤0,001 %
Iron (Fe) ≤0,001 %
For Laboratory, Research or Manufacturing Use

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR