Kaliumhydroksid 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Danger

Synonymer: Potash caustic, Caustic potash

31300.291EA 635 NOK
31300.291
Kaliumhydroksid 1 mol/l (1 N) in aqueous solution, AVS TITRINORM volumetric solution
Kaliumhydroksid
Formel: KOH
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003553
CAS-nummer: 1310-58-3
EINECS: 215-181-3
FN: 1814
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR