Di-n-octyl phthalate, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

31301-250MG
31301-250MGEA 550 NOK
31301-250MG
Di-n-octyl phthalate, Supelco®
Di-n-octyl phthalate
Formel: C₂₄H₃₈O₄
MW: 390.56 g/mol
MDL nummer: MFCD00015292
CAS-nummer: 117-84-0
EINECS: 204-214-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9022b472468bcde33bb0ef4b73293d5375b9078&vid=5340e7aa43a6a7ed8764b75a9b214262850d9cf0&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR