Ortofosforsyre 85%, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

3132-46EA 1650 NOK
3132-46
Ortofosforsyre 85%, AR®, Macron Fine Chemicals™
Ortofosforsyre
Formel: H₃PO₄
MDL nummer: MFCD00011340
CAS-nummer: 7664-38-2
FN: 1805
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear, Colourless, Syrupy Liquid
Assay ≥84.5%
Reducing substances (as H₃PO₃) ≤0.1%
Nitrate (NO₃) ≤0.001%
Chloride (Cl) ≤0.001%
Sulphate (SO₄) ≤0.01%
Calcium and Magnesium (as Ca) ≤0.02%
Arsenic (As) ≤0.0002%
Iron (Fe) ≤0.005%
Heavy Metals (as Pb) ≤0.002%
For Laboratory, Research or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR