Folin-Ciocalteu's reagent for analysis of phenols

Leverandør: VWR Chemicals
Warning

Synonymer: Folin & Ciocalteu’s phenol reagent

31360.264EA 1275 NOK
31360.264
Folin-Ciocalteu's reagent for analysis of phenols
Folin-Ciocalteu's reagent
Tetthet: 1.22 g/cm³ (20 °C)
MDL nummer: MFCD00132625
FN: 3264
ADR: 8 III

Bestill nå

Spesifikasjon Testresultater

Identification Passes test
Sensitivity to phenol Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR