Kaliumdikromat 1/6 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric

Leverandør: VWR Chemicals
Danger

31396.291EA 459 NOK
31396.291
Kaliumdikromat 1/6 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Kaliumdikromat
Formel: K₂Cr₂O₇
MW: 294.18 g/mol
Kokepunkt: 500
Smeltepunkt: 398
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011367
CAS-nummer: 7778-50-9
EINECS: 231-906-6
FN: 3287
ADR: 6.1 III
Merck Indeks: 12,07790

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.1657 - 0.1663 mol/l
Appearance Clear orange liquid
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR