di-Natriumtetraborat dekahydrat ≥99.5%, Puriss. p.a. ISO, ACS buffer substance, Fluka™

Leverandør: Honeywell Chemicals
Danger

Synonymer: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

31457-100G 31457-1KG 31457-500G 31457-5KG
31457-100GEA 865 NOK
31457-100G 31457-1KG 31457-500G 31457-5KG
di-Natriumtetraborat dekahydrat ≥99.5%, Puriss. p.a. ISO, ACS buffer substance, Fluka™
di-Natriumtetraborat dekahydrat
Formel: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Kokepunkt: 320
Smeltepunkt: 742 °C (anhy.) ≥ 62 °C (–H₂O of crystall.)
Tetthet: 1.7 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149193
CAS-nummer: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Indeks: 13,08662

Bestill nå

Spesifikasjon Testresultater

Assay 99.5 - 105.0 %
Water insoluble matter Max. 0.001 %
pH (5 %, 20°C) 9.0 - 9.6
pH (0.01 m, 25°C) 9.15 - 9.20
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR