Indikator Strips, Non-Bleeding
Leverandør: VWR Chemicals
315052JEA 200 NOK
315052J
Indikator Strips, Non-Bleeding
Testpapir pH-indikatorstrips
Non bleeding special indicator dyes that are covalently bound onto the reagent paper and then adhered to plastic strips.