Supelco® Organic Reference Standard, Anthracene

Leverandør: Merck

31581-250MG
31581-250MGEA 1225 NOK
31581-250MG
Supelco® Organic Reference Standard, Anthracene
Standarder Organiske standarder

  • GHS signal: GHS07, GHS09


NB: P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P391

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce86ad55635d42c326a54ca9dcf73408404e0a3&vid=8ce86ad55635d42c326a54ca9dcf73408404e0a3&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) Anthraxcene; Paranaphthalene
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 120-12-7
MDL No. MFCD00001240
Molekylvekt 178,23
Smeltepunkt 210...215 °C (lit.)
kokepunkt 340 °C (lit.)
Hardhetsklasse beskrivelse H319 - H410
Flammepunkt 121,0 °C - closed cup
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR