Natriumhydroksid 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

31630.303EA 1050 NOK
31630.303
Natriumhydroksid 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Natriumhydroksid
Formel: NaOH
Tetthet: 1 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR