Supelco® PESTANAL® Organic Reference Standard, Fenhexamid

Leverandør: Merck

31713-100MG
31713-100MGEA 1250 NOK
31713-100MG
Supelco® PESTANAL® Organic Reference Standard, Fenhexamid
Standarder Organiske standarder

  • GHS signal: GHS09


NB: P273 - P391 - P501

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8a1d7fcf67197886edc520dcc05f8ebec8ae44c&vid=af29749764d554202c294ff07fcaa457d59efc36&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 126833-17-8
MDL No. MFCD03095700
Formel C₁₄H₁₇Cl₂NO₂
Molekylvekt 302,20
Hardhetsklasse beskrivelse H411
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR