Kongorødt 1% in aqueous solution

Leverandør: VWR Chemicals

31727.295EA 2275 NOK
31727.295
Kongorødt 1% in aqueous solution
Kongorødt

  • Transition range: pH 3,0 - 5,2

Formel: C₃₂H₂₂N₆Na₂O₆S₂
Kokepunkt: ~100 °C (1013 hPa)
Tetthet: 1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004028
CAS-nummer: 573-58-0
EINECS: 209-358-4

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red liquid
Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR