Natriumhydroksid 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution

Leverandør: VWR Chemicals
Warning

31951.290EA 390 NOK
31951.290 31951.370 31951.404
Natriumhydroksid 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Natriumhydroksid
Formel: NaOH
Kokepunkt: >105 °C (1013 hPa)
Tetthet: 1,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR