Natriumhydroksid 0.25 mol/l (0.25 N) in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

Synonymer: Caustic soda

31956.296EA 385 NOK
31956.296 31956.410
Natriumhydroksid 0.25 mol/l (0.25 N) in aqueous solution, AVS TITRINORM volumetric solution
Natriumhydroksid
Formel: NaOH
Kokepunkt: >105 °C (1013 hPa)
Tetthet: 1,02 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.2495 - 0.2505 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR