Ammoniumtiocyanat 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric

Leverandør: VWR Chemicals

Synonymer: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

31960.291EA 1450 NOK
31960.291
Ammoniumtiocyanat 1 mol/l (1 N) aqueous solution, AVS TITRINORM Volumetric
Ammoniumtiocyanat
Formel: NH₄SCN
MW: 76.12 g/mol
Smeltepunkt: 150
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011428
CAS-nummer: 1762-95-4
EINECS: 217-175-6
Merck Indeks: 13,00564

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR