Saltsyre 0.2 mol/l (0.2 N), AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

31983.290EA 435 NOK
31983.290 31983.370
Saltsyre 0.2 mol/l (0.2 N), AVS TITRINORM volumetric solution
Saltsyre
Formel: HCl
Tetthet: 1,004 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.1996 - 0.2004 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR